siRNA Details
Gene Symbol: Esrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107423.1 | NP_001100893.1 | 1 | 386 | ||
XM_006255496.2 | XP_006255558.1 | 1 | 259 | |||
GenBank mRNA | BC166804.1 | - | 1 | 247 | 7938923 |
