siRNA Details
Gene Symbol: Mon1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107433.1 | NP_001100903.1 | 2 | 554 | ||
XM_006255674.2 | XP_006255736.1 | 4 | 1075 | |||
XM_006255675.2 | XP_006255737.1 | 4 | 889 | |||
XM_008772515.1 | XP_008770737.1 | 3 | 1104 |
