siRNA Details
Gene Symbol: Mesp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107531.1 | NP_001101001.1 | 1 | 782 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mesp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107531.1 | NP_001101001.1 | 1 | 782 |