siRNA Details
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100582.1 | NP_001094052.1 | 16 | 1666 | ||
XM_006229953.2 | XP_006230015.1 | 15 | 1609 | |||
XM_006229955.2 | XP_006230017.1 | 12 | 1970 | |||
XM_008759733.1 | XP_008757955.1 | 17 | 1671 | |||
XM_008759734.1 | XP_008757956.1 | 18 | 2029 | |||
XM_008759735.1 | XP_008757957.1 | 16 | 1792 | |||
XM_008759736.1 | XP_008757958.1 | 14 | 2029 | |||
GenBank mRNA | AY754026.1 | - | 11 | 884 |
