siRNA Details
Gene Symbol: Ppfibp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100582.1 | NP_001094052.1 | 21 | 2321 | ||
XM_006229953.2 | XP_006230015.1 | 20 | 2264 | |||
XM_006229955.2 | XP_006230017.1 | 17 | 2625 | |||
XM_008759733.1 | XP_008757955.1 | 22 | 2326 | |||
XM_008759734.1 | XP_008757956.1 | 23 | 2684 | |||
XM_008759735.1 | XP_008757957.1 | 21 | 2447 | |||
XM_008759736.1 | XP_008757958.1 | 19 | 2684 | |||
GenBank mRNA | AY754026.1 | - | 16 | 1539 |
