siRNA Details
Gene Symbol: Fbxl19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107552.1 | NP_001101022.1 | 8 | 1475 | ||
XM_006230276.2 | XP_006230338.1 | 11 | 2333 | |||
XM_006230277.2 | XP_006230339.1 | 11 | 2204 | |||
XM_006230278.2 | XP_006230340.1 | 10 | 2114 | |||
XM_006230279.2 | XP_006230341.1 | 10 | 1985 | |||
XM_006230280.2 | XP_006230342.1 | 11 | 2217 |
