siRNA Details
Gene Symbol: LOC102554620 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773895.1 | XP_008772117.1 | 1721 | |||
XM_008773897.1 | XP_008772119.1 | 2212 |
Gene Symbol: Vwce | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271311.1 | NP_001258240.1 | 15, 16 | 1859 |
