siRNA Details
Gene Symbol: Ms4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107578.1 | NP_001101048.1 | 4 | 500 | ||
XM_006231089.2 | XP_006231151.1 | 5 | 672 | |||
GenBank mRNA | BC098745.1 | - | 5 | 563 | ||
FQ225563.1 | - | 5 | 623 | |||
FQ235337.1 | - | 4 | 437 |
