siRNA Details
Gene Symbol: Vps13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100975.2 | NP_001094445.2 | 52 | 7365 | ||
GenBank mRNA | AY325251.1 | - | 8 | 1449 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Vps13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100975.2 | NP_001094445.2 | 52 | 7365 | ||
GenBank mRNA | AY325251.1 | - | 8 | 1449 |