siRNA Details
Gene Symbol: Vps13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100975.2 | NP_001094445.2 | 47 | 6466 | ||
XM_008760289.1 | XP_008758511.1 | 48 | 6765 | |||
GenBank mRNA | AY325251.1 | - | 2 | 256 |
