siRNA Details
Gene Symbol: Gldc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107583.1 | NP_001101053.1 | 17 | 2317 | ||
XM_006231210.2 | XP_006231272.1 | 21 | 2718 | |||
GenBank mRNA | FQ209556.1 | - | 3 | 230 |
