siRNA Details
Gene Symbol: Fam122a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014029.1 | NP_001014051.1 | 1 | 639 | ||
GenBank mRNA | BC078922.1 | - | 1 | 639 | 7106028 |

More Information
Other Species Targeted

