siRNA Details
Gene Symbol: Kank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037197.2 | NP_001032274.2 | 5, 6 | 3083 | ||
XM_006231212.2 | XP_006231274.1 | 3, 4 | 4026 | |||
XM_006231213.2 | XP_006231275.1 | 3, 4 | 5707 | |||
XM_006231214.2 | XP_006231276.1 | 5, 6 | 3200 | |||
XM_006231215.2 | XP_006231277.1 | 4, 5 | 3111 | |||
XM_006231216.2 | XP_006231278.1 | 5, 6 | 5634 | |||
XM_006231218.2 | XP_006231280.1 | 3, 4 | 4026 | |||
XM_006231219.1 | XP_006231281.1 | 3, 4 | 3024 | |||
XM_006231220.1 | XP_006231282.1 | 3, 4 | 2947 | |||
XM_008760305.1 | XP_008758527.1 | 5, 6 | 3217 | |||
GenBank mRNA | BC107446.1 | - | 2, 3 | 292 |
