siRNA Details
Gene Symbol: LOC100912537 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749109.3 | XP_003749157.1 | 1598 |
Gene Symbol: Tctn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_220007.8 | XP_220007.4 | 13 | 1598 |
