siRNA Details
Gene Symbol: Rtkn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107625.1 | NP_001101095.1 | 8 | 1097 | ||
XM_006256367.2 | XP_006256429.1 | 9 | 1178 | |||
XM_008772883.1 | XP_008771105.1 | 9 | 2647 | |||
XM_008772884.1 | XP_008771106.1 | 8 | 810 |
