siRNA Details
Gene Symbol: Ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047886.1 | NP_001041351.1 | 14, 15 | 1281 | ||
XM_006232624.2 | XP_006232686.1 | 13, 14 | 1136 | |||
GenBank mRNA | AY325142.1 | - | 14, 15 | 1281 | ||
BC167105.1 | - | 13, 14 | 1036 | 9102039 |
