siRNA Details
Gene Symbol: Trim46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107691.1 | NP_001101161.1 | 2 | 333 | ||
XM_008761170.1 | XP_008759392.1 | 2 | 450 | |||
XM_008761171.1 | XP_008759393.1 | 2 | 297 | |||
XM_008761172.1 | XP_008759394.1 | 2 | 296 | |||
XM_008761173.1 | XP_008759395.1 | 2 | 297 |

More Information
Other Species Targeted

Gene Ontology Categories:
