siRNA Details
Gene Symbol: Dclk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009691.3 | NP_001009691.3 | 1 | 629 | ||
NM_001195832.1 | NP_001182761.1 | 1 | 629 | |||
XM_006232679.2 | XP_006232741.1 | 1 | 799 | |||
XM_006232680.2 | XP_006232742.1 | 1 | 799 | |||
XM_006232681.2 | XP_006232743.1 | 1 | 798 | |||
XM_006232682.2 | XP_006232744.1 | 1 | 798 | |||
XM_006232684.2 | XP_006232746.1 | 1 | 799 | |||
XM_006232685.2 | XP_006232747.1 | 1 | 797 | |||
XM_008761176.1 | XP_008759398.1 | 1 | 800 | |||
XM_008761178.1 | XP_008759400.1 | 1 | 797 | |||
GenBank mRNA | AY673997.1 | - | 1 | 21 | ||
AY673998.1 | - | 1 | 101 | |||
AY674000.1 | - | 1 | 21 |
