siRNA Details
Gene Symbol: Fnbp1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039609.1 | NP_001034698.1 | 1335 | |||
GenBank mRNA | AB250295.1 | - | 1335 | |||
AB250296.1 | - | 1161 |

More Information
Other Species Targeted

Gene Ontology Categories:
