siRNA Details
Gene Symbol: Tifa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014044.1 | NP_001014066.1 | 3 | 527 | ||
XM_006233289.2 | XP_006233351.1 | 3 | 717 | |||
XM_006233290.2 | XP_006233352.1 | 3 | 694 | |||
XM_008761463.1 | XP_008759685.1 | 3 | 866 | |||
XM_008761464.1 | XP_008759686.1 | 3 | 626 | |||
GenBank mRNA | AY325204.1 | - | 970 | |||
BC083765.1 | - | 3 | 527 | 7193165 |
