siRNA Details
Gene Symbol: Zgrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233293.2 | XP_006233355.1 | 7 | 3310 | ||
XM_006233294.2 | XP_006233356.1 | 7 | 2706 | |||
XM_006233296.2 | XP_006233358.1 | 6 | 3182 | |||
XM_006233297.2 | XP_006233359.1 | 2 | 1799 | |||
XM_008761479.1 | XP_008759701.1 | 6 | 3201 |
