siRNA Details
Gene Symbol: Gbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233424.2 | XP_006233486.1 | 4, 5 | 810 | ||
XM_227762.8 | XP_227762.6 | 4, 5 | 812 | |||
GenBank mRNA | FQ225190.1 | - | 4, 5 | 588 |
Gene Symbol: LOC100911495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008761539.1 | XP_008759761.1 | 804 |
