siRNA Details
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107755.1 | NP_001101225.1 | 2, 3 | 308 | ||
XM_008762061.1 | XP_008760283.1 | 2, 3 | 425 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_173749, BC031841, BC057685, AK031254, BB620582, and others.
