siRNA Details
Gene Symbol: Casc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170594.1 | NP_001164065.1 | 17 | 5665 | ||
XM_008762095.1 | XP_008760317.1 | 20 | 5851 | |||
XM_008762096.1 | XP_008760318.1 | 20 | 6081 | |||
XM_008762097.1 | XP_008760319.1 | 20 | 5894 | |||
XM_008762098.1 | XP_008760320.1 | 18 | 5764 | |||
XM_008762099.1 | XP_008760321.1 | 20 | 5838 |
