siRNA Details
Gene Symbol: Soga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235395.2 | XP_006235457.2 | 14 | 3964 | ||
GenBank mRNA | AB190506.1 | - | 1 | 23 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Soga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235395.2 | XP_006235457.2 | 14 | 3964 | ||
GenBank mRNA | AB190506.1 | - | 1 | 23 |