siRNA Details
Gene Symbol: Gdap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107798.2 | NP_001101268.1 | 5 | 674 | ||
GenBank mRNA | BC168159.1 | - | 5 | 674 | 8364600 | |
FQ212886.1 | - | 5 | 731 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gdap1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107798.2 | NP_001101268.1 | 5 | 674 | ||
GenBank mRNA | BC168159.1 | - | 5 | 674 | 8364600 | |
FQ212886.1 | - | 5 | 731 |