siRNA Details
Gene Symbol: Stkld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107822.1 | NP_001101292.1 | 18, 19 | 1945 | ||
XM_006233785.2 | XP_006233847.1 | 18, 19 | 2156 | |||
XM_008761643.1 | XP_008759865.1 | 17, 18 | 2333 | |||
XM_008761644.1 | XP_008759866.1 | 16, 17 | 2204 | |||
XM_008761645.1 | XP_008759867.1 | 15, 16 | 2075 | |||
GenBank mRNA | BC161886.1 | - | 18, 19 | 1928 | 7133966 |
