siRNA Details
Gene Symbol: Gfi1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107823.1 | NP_001101293.1 | 7 | 1088 | ||
XM_006233789.1 | XP_006233851.1 | 7 | 1226 | |||
XM_008761646.1 | XP_008759868.1 | 7 | 1274 | |||
XM_008761647.1 | XP_008759869.1 | 7 | 4548 | |||
XM_008761648.1 | XP_008759870.1 | 8 | 1438 | |||
XM_008761649.1 | XP_008759871.1 | 7 | 1348 | |||
XM_008761650.1 | XP_008759872.1 | 7 | 1826 | |||
XM_008761651.1 | XP_008759873.1 | 7 | 1175 | |||
GenBank mRNA | BC158685.1 | - | 7 | 1103 | 8374780 |
