siRNA Details
Gene Symbol: Usp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107827.3 | NP_001101297.1 | 18 | 1959 | ||
XM_006233916.2 | XP_006233978.1 | 18 | 3094 | |||
XM_008761666.1 | XP_008759888.1 | 16 | 2068 | |||
XM_008761667.1 | XP_008759889.1 | 16 | 2026 |

More Information
Other Species Targeted

Gene Ontology Categories:
