siRNA Details
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109666.1 | NP_001103136.1 | 9 | 1367 | ||
XM_006235929.2 | XP_006235991.1 | 10 | 1337 | |||
XM_006235930.2 | XP_006235992.1 | 10 | 1499 | |||
XM_006235931.2 | XP_006235993.1 | 10 | 1334 | |||
XM_006235932.2 | XP_006235994.1 | 10 | 1328 | |||
XM_006235933.2 | XP_006235995.1 | 10 | 1325 | |||
GenBank mRNA | BC129119.1 | - | 9 | 1234 | 7115823 |
