siRNA Details
Gene Symbol: Dbf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191748.1 | NP_001178677.1 | 565 |
Gene Symbol: LOC103690019 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762747.1 | XP_008760969.1 | 4 | 646 |
