siRNA Details
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107848.1 | NP_001101318.1 | 4 | 664 | ||
XM_006257072.2 | XP_006257134.1 | 3 | 1105 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ophn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107848.1 | NP_001101318.1 | 4 | 664 | ||
XM_006257072.2 | XP_006257134.1 | 3 | 1105 |