siRNA Details
Gene Symbol: Nrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100708.1 | NP_001094178.1 | 3 | 276 | ||
XM_006236198.2 | XP_006236260.1 | 3 | 324 | |||
XM_006236200.2 | XP_006236262.1 | 3 | 324 | |||
XM_008762774.1 | XP_008760996.1 | 3 | 322 | |||
GenBank mRNA | BC161956.1 | - | 3 | 276 | 9027258 |
