siRNA Details
Gene Symbol: RGD1306746 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107872.2 | NP_001101342.2 | 5 | 1309 | ||
XM_008763070.1 | XP_008761292.1 | 5 | 1587 | |||
XM_008763071.1 | XP_008761293.1 | 5 | 1359 | |||
XM_008763072.1 | XP_008761294.1 | 5 | 1709 | |||
XM_008763073.1 | XP_008761295.1 | 5 | 1534 | |||
XM_008763074.1 | XP_008761296.1 | 5 | 3215 | |||
GenBank mRNA | BC166726.1 | - | 5 | 1309 | 7933964 |
