siRNA Details
Gene Symbol: Lrig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763142.1 | XP_008761364.1 | 5 | 1364 | ||
XM_232237.8 | XP_232237.6 | 5 | 576 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrig1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763142.1 | XP_008761364.1 | 5 | 1364 | ||
XM_232237.8 | XP_232237.6 | 5 | 576 |