siRNA Details
Gene Symbol: Rasgef1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237211.2 | XP_006237273.1 | 5 | 602 | ||
XM_008763269.1 | XP_008761491.1 | 5 | 738 | |||
XM_232315.7 | XP_232315.6 | 5 | 764 |
