siRNA Details
Gene Symbol: Il17ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107883.2 | NP_001101353.2 | 13 | 2693 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Il17ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107883.2 | NP_001101353.2 | 13 | 2693 |