siRNA Details
Gene Symbol: Cops7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047098.2 | NP_001040563.2 | 6, 7 | 787 | ||
NM_001164091.1 | NP_001157563.1 | 6, 7 | 803 | |||
XM_006237367.1 | XP_006237429.1 | 6, 7 | 811 | |||
XM_006237368.2 | XP_006237430.1 | 6, 7 | 997 | |||
GenBank mRNA | AB259153.1 | - | 5, 6 | 515 | ||
BI296165.1 | - | 2, 3 | 475 | |||
FQ215513.1 | - | 6, 7 | 811 |
