siRNA Details
Gene Symbol: Tram1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007701.1 | NP_001007702.1 | 1088 | |||
GenBank mRNA | AY387055.1 | - | 971 | |||
BC083851.1 | - | 1088 | 7123274 | |||
FQ223598.1 | - | 960 |
