siRNA Details
Gene Symbol: Cpa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107900.1 | NP_001101370.1 | 9 | 934 | ||
XM_008763496.1 | XP_008761718.1 | 9 | 1025 |

More Information
Other Species Targeted

Gene Ontology Categories:
