siRNA Details
Gene Symbol: Asph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237855.2 | XP_006237917.1 | 4 | 749 | ||
XM_006237858.2 | XP_006237920.1 | 3 | 702 | |||
XM_006237859.2 | XP_006237921.1 | 4 | 2033 | |||
XM_006237860.2 | XP_006237922.1 | 3 | 1987 | |||
XM_006237861.2 | XP_006237923.1 | 3 | 704 | |||
XM_006237862.2 | XP_006237924.1 | 4 | 2034 | |||
XM_006237863.2 | XP_006237925.1 | 3 | 702 | |||
XM_006237864.2 | XP_006237926.1 | 3 | 1986 | |||
XM_008763567.1 | XP_008761789.1 | 4 | 749 | |||
XM_008763568.1 | XP_008761790.1 | 3 | 703 | |||
XM_008763569.1 | XP_008761791.1 | 4 | 750 | |||
XM_008763570.1 | XP_008761792.1 | 4 | 2034 | |||
XM_008763571.1 | XP_008761793.1 | 4 | 750 | |||
XM_008763572.1 | XP_008761794.1 | 3 | 703 | |||
XM_008763573.1 | XP_008761795.1 | 3 | 1988 | |||
XM_008763574.1 | XP_008761796.1 | 4 | 2034 | |||
XM_008763575.1 | XP_008761797.1 | 4 | 753 | |||
GenBank mRNA | BC085901.1 | - | 3 | 280 | 7193538 |
