siRNA Details
Gene Symbol: Asph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237863.2 | XP_006237925.1 | 5 | 1105 | ||
XM_006237864.2 | XP_006237926.1 | 5 | 2389 | |||
XM_008763574.1 | XP_008761796.1 | 6 | 2437 | |||
GenBank mRNA | BC085901.1 | - | 5 | 683 | 7193538 | |
FQ217288.1 | - | 1 | 79 |
