siRNA Details
Gene Symbol: Casp8ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107921.1 | NP_001101391.1 | 8 | 4553 | ||
XM_006237973.2 | XP_006238035.1 | 9 | 4453 | |||
XM_006237974.2 | XP_006238036.1 | 9 | 4629 | |||
XM_006237975.2 | XP_006238037.1 | 8 | 4383 | |||
XM_008763620.1 | XP_008761842.1 | 8 | 5547 |
