siRNA Details
Gene Symbol: Nfx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024784.2 | NP_001019955.2 | 3 | 1255 | ||
XM_008763623.1 | XP_008761845.1 | 3 | 1473 | |||
GenBank mRNA | BC098037.1 | - | 3 | 1161 | 7456610 |
