siRNA Details
Gene Symbol: Rhox4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024889.1 | NP_001020060.1 | 4 | 570 | ||
GenBank mRNA | DQ058652.1 | - | 4 | 570 |
Gene Symbol: LOC103689929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773509.1 | XP_008771731.1 | 4 | 570 |
