siRNA Details
Gene Symbol: Zbtb17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012105.1 | NP_001012105.1 | 3 | 327 | ||
XM_006239303.1 | XP_006239365.1 | 3 | 322 | |||
XM_008764253.1 | XP_008762475.1 | 2 | 233 | |||
GenBank mRNA | BC083577.1 | - | 3 | 327 | 7113925 |

More Information
Other Species Targeted

Gene Ontology Categories:
