siRNA Details
Gene Symbol: Nphp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037650.2 | NP_001032739.2 | 21 | 3320 | ||
XM_006239464.2 | XP_006239526.1 | 22 | 11067 | |||
XM_006239465.2 | XP_006239527.1 | 21 | 3107 | |||
XM_006239467.2 | XP_006239529.1 | 22 | 4452 | |||
XM_008764358.1 | XP_008762580.1 | 23 | 4582 | |||
XM_008764359.1 | XP_008762581.1 | 21 | 10992 | |||
GenBank mRNA | BC107905.1 | - | 1 | 392 |
