siRNA Details
Gene Symbol: Clec14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014077.1 | NP_001014099.1 | 1 | 1180 | ||
GenBank mRNA | BC081897.1 | - | 1 | 1180 | 7110446 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Clec14a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014077.1 | NP_001014099.1 | 1 | 1180 | ||
GenBank mRNA | BC081897.1 | - | 1 | 1180 | 7110446 |