siRNA Details
Gene Symbol: Gpatch2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134559.1 | NP_001128031.1 | 2, 3 | 734 | ||
XM_008764793.1 | XP_008763015.1 | 2, 3 | 779 | |||
XM_008764794.1 | XP_008763016.1 | 2, 3 | 774 | |||
GenBank mRNA | FQ212324.1 | - | 2, 3 | 742 |
