siRNA Details
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005888.1 | NP_001005888.1 | 1, 2 | 199 | ||
GenBank mRNA | AY386370.1 | - | 1, 2 | 199 | ||
FQ223346.1 | - | 1, 2 | 218 | |||
FQ228993.1 | - | 1, 2 | 218 |
